Assay Details
| Gene Symbol: ZNF711 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014111881.2 | XP_013967356.1 | 7 | 1389 | 90 | |
| XM_022415877.1 | XP_022271585.1 | 5 | 1016 | 90 | ||
| XM_022415878.1 | XP_022271586.1 | 7 | 1394 | 90 | ||
| XM_022415879.1 | XP_022271587.1 | 6 | 1261 | 90 | ||
| XM_022415880.1 | XP_022271588.1 | 6 | 1132 | 90 | ||
| XM_022415882.1 | XP_022271590.1 | 5 | 1016 | 90 | ||
| XM_022415883.1 | XP_022271591.1 | 5 | 1016 | 90 | ||
| XM_022415884.1 | XP_022271592.1 | 5 | 1016 | 90 | ||
| XM_022415885.1 | XP_022271593.1 | 3 | 419 | 90 | ||
More Information
Set Membership: |
|

ZNF711
Genomic Map