Assay Details
| Gene Symbol: VPS37B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005636144.3 | XP_005636201.1 | 346 | 161 | ||
| XM_014107872.2 | XP_013963347.1 | 484 | 161 | |||
| XM_534655.6 | XP_534655.2 | 413 | 161 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

VPS37B
Genomic Map