Assay Details
| Gene Symbol: USP44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005629240.2 | XP_005629297.1 | 4 | 506 | 151 | |
| XM_005629243.2 | XP_005629300.1 | 3 | 439 | 151 | ||
| XM_005629244.2 | XP_005629301.1 | 3 | 360 | 151 | ||
| XM_005629247.3 | XP_005629304.1 | 2 | 449 | 151 | ||
| XM_005629248.3 | XP_005629305.1 | 2 | 288 | 151 | ||
| XM_022403339.1 | XP_022259047.1 | 3 | 493 | 151 | ||
| XM_022403340.1 | XP_022259048.1 | 4 | 567 | 151 | ||
| XM_022403341.1 | XP_022259049.1 | 3 | 424 | 151 | ||
| XM_532654.5 | XP_532654.1 | 3 | 413 | 151 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

USP44
Genomic Map