Assay Details
Gene Symbol: AGBL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022427936.1 | XP_022283644.1 | 11 - 12 | 1540 | 163 | |
XM_022427938.1 | XP_022283646.1 | 11 - 12 | 1540 | 163 | ||
XM_022427939.1 | XP_022283647.1 | 11 - 12 | 1540 | 163 | ||
XM_022427940.1 | XP_022283648.1 | 11 - 12 | 1540 | 163 | ||
XM_022427941.1 | XP_022283649.1 | 9 - 10 | 1295 | 163 | ||
XM_022427942.1 | XP_022283650.1 | 9 - 10 | 1314 | 163 | ||
XM_022427943.1 | XP_022283651.1 | 9 - 10 | 1305 | 163 | ||
XM_022427945.1 | XP_022283653.1 | 9 - 10 | 1351 | 163 | ||
XM_022427947.1 | XP_022283655.1 | 9 - 10 | 1021 | 163 | ||
XM_022427949.1 | XP_022283657.1 | 6 - 7 | 1857 | 163 | ||
XM_022427950.1 | XP_022283658.1 | 6 - 7 | 3440 | 163 | ||
XM_022427951.1 | XP_022283659.1 | 7 - 8 | 3315 | 163 | ||
XM_022427952.1 | XP_022283660.1 | 6 - 7 | 5389 | 163 | ||
XM_847536.4 | XP_852629.2 | 9 - 10 | 1021 | 163 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
