Assay Details
| Gene Symbol: PDGFB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003383.1 | NP_001003383.1 | 3 | 179 | 149 | |
| XM_005625680.3 | XP_005625737.1 | 3 | 240 | 149 | ||
| XM_005625682.3 | XP_005625739.1 | 3 | 240 | 149 | ||
| XM_005625683.3 | XP_005625740.1 | 3 | 240 | 149 | ||
| XM_022423987.1 | XP_022279695.1 | 3 | 272 | 149 | ||
| GenBank mRNA | AY553983.1 | - | 3 | 179 | 149 | |
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PDGFB
Genomic Map