Assay Details
| Gene Symbol: ATOH7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005618942.3 | XP_005618999.1 | 1 | 445 | 164 | |
| XM_022417486.1 | XP_022273194.1 | 1 | 445 | 164 | ||
More Information
Set Membership: |
|

ATOH7
Genomic Map