Assay Details
| Gene Symbol: ARRDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003435333.4 | XP_003435381.1 | 5 | 621 | 237 | |
| XM_022423503.1 | XP_022279211.1 | 6 | 684 | 237 | ||
| XM_022423504.1 | XP_022279212.1 | 5 | 515 | 237 | ||
More Information
Set Membership: |
|

ARRDC1
Genomic Map