Assay Details
| Gene Symbol: DNAJC18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022408276.1 | XP_022263984.1 | 211 | 166 | ||
| XM_022408283.1 | XP_022263991.1 | 211 | 166 | |||
| XM_022408290.1 | XP_022263998.1 | 211 | 166 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

DNAJC18
Genomic Map