Assay Details
| Gene Symbol: LIX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005618066.3 | XP_005618123.1 | 522 | 163 | ||
| XM_022416698.1 | XP_022272406.1 | 291 | 163 | |||
| XM_022416699.1 | XP_022272407.1 | 578 | 163 | |||
| XM_546011.6 | XP_546011.2 | 518 | 163 | |||
More Information
Set Membership: |
|

LIX1
Genomic Map