Assay Details
| Gene Symbol: NYAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005621044.3 | XP_005621101.1 | 261 | 151 | ||
| XM_005621045.3 | XP_005621102.1 | 825 | 151 | |||
| XM_005621046.3 | XP_005621103.1 | 261 | 151 | |||
| XM_022419642.1 | XP_022275350.1 | 332 | 151 | |||
| XM_546956.6 | XP_546956.3 | 409 | 151 | |||
More Information
Set Membership: |
|

NYAP1
Genomic Map