Assay Details
| Gene Symbol: LSM14A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005616759.3 | XP_005616816.1 | 3 - 4 | 384 | 72 | |
| XM_005616760.3 | XP_005616817.1 | 3 - 4 | 384 | 72 | ||
| XM_005616761.2 | XP_005616818.1 | 3 - 4 | 360 | 72 | ||
| XM_014120759.2 | XP_013976234.1 | 3 - 4 | 312 | 72 | ||
| XM_022426175.1 | XP_022281883.1 | 3 - 4 | 384 | 72 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

LSM14A
Genomic Map