Assay Details
| Gene Symbol: KLK13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003638808.4 | XP_003638856.1 | 5 - 6 | 1786 | 101 | |
| XM_005616251.2 | XP_005616308.1 | 4 - 5 | 610 | 101 | ||
| XM_022423157.1 | XP_022278865.1 | 5 - 6 | 1063 | 101 | ||
| XM_022423168.1 | XP_022278876.1 | 3 - 4 | 604 | 101 | ||
| XM_022423169.1 | XP_022278877.1 | 3 - 4 | 1322 | 101 | ||
| GenBank mRNA | KJ831042.1 | - | 4 - 5 | 655 | 101 | |
| KJ831043.1 | - | 2 - 3 | 196 | 101 | ||
More Information
Set Membership: |
|

KLK13
Genomic Map