Assay Details
| Gene Symbol: ZDHHC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005632643.3 | XP_005632700.1 | 4 - 5 | 789 | 143 | |
| XM_005632644.3 | XP_005632701.1 | 4 - 5 | 900 | 143 | ||
| XM_022406805.1 | XP_022262513.1 | 4 - 5 | 898 | 143 | ||
| XM_022406806.1 | XP_022262514.1 | 4 - 5 | 898 | 143 | ||
| XM_022406807.1 | XP_022262515.1 | 4 - 5 | 897 | 143 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

ZDHHC3
Genomic Map