Assay Details
| Gene Symbol: SLC22A12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284473.1 | NP_001271402.1 | 4 - 5 | 828 | 169 | |
| XM_005631536.3 | XP_005631593.1 | 4 - 5 | 1210 | 169 | ||
| XM_014120874.2 | XP_013976349.1 | 4 - 5 | 1210 | 169 | ||
| GenBank mRNA | AB738915.1 | - | 4 - 5 | 828 | 169 | |
More Information
Set Membership: |
|

SLC22A12
Genomic Map