Assay Details
| Gene Symbol: CNEP1R1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005617579.3 | XP_005617636.1 | 4 - 5 | 407 | 113 | |
| XM_022411517.1 | XP_022267225.1 | 3 - 4 | 469 | 113 | ||
More Information
Set Membership: |
|

CNEP1R1
Genomic Map