Assay Details
| Gene Symbol: MOSPD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005641844.3 | XP_005641901.1 | 5 - 6 | 926 | 142 | |
| XM_538179.6 | XP_538179.1 | 4 - 5 | 709 | 142 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

MOSPD1
Genomic Map