Assay Details
| Gene Symbol: BCL9L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014113219.2 | XP_013968694.1 | 4586 | 133 | ||
| XM_022418222.1 | XP_022273930.1 | 4528 | 133 | |||
| XM_022418223.1 | XP_022273931.1 | 4604 | 133 | |||
More Information
Set Membership: |
|

BCL9L
Genomic Map