Assay Details
| Gene Symbol: SPRY4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005617324.3 | XP_005617381.1 | 291 | 160 | ||
| XM_005617325.3 | XP_005617382.1 | 219 | 160 | |||
| XM_022408905.1 | XP_022264613.1 | 3224 | 160 | |||
More Information
Set Membership: |
|

SPRY4
Genomic Map