Assay Details
| Gene Symbol: DCN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003228.1 | NP_001003228.1 | 7 - 8 | 1068 | 134 | |
| XM_005628825.3 | XP_005628882.1 | 7 - 8 | 1073 | 134 | ||
| XM_005628826.3 | XP_005628883.1 | 9 - 10 | 1204 | 134 | ||
| XM_005628828.1 | XP_005628885.1 | 7 - 8 | 1164 | 134 | ||
| XM_022427654.1 | XP_022283362.1 | 8 - 9 | 1166 | 134 | ||
| GenBank mRNA | U83141.1 | - | 7 - 8 | 1068 | 134 | |
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

DCN
Genomic Map