Assay Details
| Gene Symbol: FTH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193656.1 | NP_001180585.1 | 1 | 173 | 142 | |
| XM_022405806.1 | XP_022261514.1 | 2 | 196 | 142 | ||
| GenBank mRNA | AB175610.1 | - | 1 | 173 | 142 | |
| AB175611.1 | - | 1 | 141 | 142 | ||
| Gene Symbol: FTH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003080.1 | NP_001003080.1 | 1 | 322 | 142 | |
| GenBank mRNA | AF285177.1 | - | 1 | 322 | 142 | |
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

FTH1
Genomic Map