Assay Details
| Gene Symbol: VPS52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001048088.1 | NP_001041553.1 | 13 - 14 | 1672 | 69 | |
| XM_005627052.2 | XP_005627109.1 | 13 - 14 | 1763 | 69 | ||
More Information
Set Membership: |
|

VPS52
Genomic Map