Assay Details
| Gene Symbol: PADI6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097547.2 | NP_001091016.2 | 6 - 7 | 732 | 65 | |
| GenBank mRNA | AY682099.1 | - | 6 - 7 | 732 | 65 | |
More Information
Set Membership: |
|

PADI6
Genomic Map