Assay Details
| Gene Symbol: SLPI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113171.1 | NP_001106642.1 | 2 - 3 | 387 | 86 | |
| GenBank mRNA | AB266058.1 | - | 2 - 3 | 387 | 86 | |
More Information
Set Membership: |
|

SLPI
Genomic Map