Assay Details
| Gene Symbol: CDKN2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001146269.1 | NP_001139741.1 | 2 - 3 | 155 | 62 | |
| XM_005626410.3 | XP_005626467.1 | 1 - 2 | 656 | 62 | ||
More Information
Set Membership: |
|

CDKN2B
Genomic Map