Assay Details
| Gene Symbol: ORMDL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022414970.1 | XP_022270678.1 | 3 - 4 | 465 | 98 | |
| XM_022414971.1 | XP_022270679.1 | 3 - 4 | 657 | 98 | ||
| XM_843173.5 | XP_848266.3 | 3 - 4 | 434 | 98 | ||
More Information
Set Membership: |
|

ORMDL1
Genomic Map