Assay Details
| Gene Symbol: CYP1A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003433890.4 | XP_003433938.1 | 6 - 7 | 1434 | 66 | |
| GenBank mRNA | KP340900.1 | - | 5 - 6 | 1293 | 66 | |
| KP340901.1 | - | 5 - 6 | 1286 | 66 | ||
More Information
Set Membership: |
|

CYP1A1
Genomic Map