Assay Details
| Gene Symbol: KLK14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001165879.1 | NP_001159351.1 | 2 - 3 | 238 | 53 | |
| XM_005615760.3 | XP_005615817.1 | 2 - 3 | 1027 | 53 | ||
| GenBank mRNA | FJ825759.1 | - | 2 - 3 | 238 | 53 | |
| FJ825760.1 | - | 2 - 3 | 238 | 53 | ||
| FJ825761.1 | - | 2 - 3 | 238 | 53 | ||
| FJ825762.1 | - | 1 - 2 | 1068 | 53 | ||
| FJ825763.1 | - | 1 - 2 | 1068 | 53 | ||
| FJ825764.1 | - | 1 - 2 | 1068 | 53 | ||
More Information
Set Membership: |
|

KLK14
Genomic Map