Assay Details
| Gene Symbol: KLK14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001165879.1 | NP_001159351.1 | 4 - 5 | 617 | 80 | |
| GenBank mRNA | FJ825759.1 | - | 4 - 5 | 617 | 80 | |
| FJ825760.1 | - | 4 - 5 | 564 | 80 | ||
| FJ825762.1 | - | 3 - 4 | 1447 | 80 | ||
| FJ825763.1 | - | 3 - 4 | 1394 | 80 | ||
More Information
Set Membership: |
|

KLK14
Genomic Map