Assay Details
Gene Symbol: KCNA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022407947.1 | XP_022263655.1 | 1 | 3811 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
