Assay Details
| Gene Symbol: PALM2AKAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204899.1 | NP_001191828.1 | 1 - 2 | 39 | 68 | |
| XM_005626411.3 | XP_005626468.2 | 2 - 3 | 107 | 68 | ||
| XM_005626413.3 | XP_005626470.2 | 2 - 3 | 107 | 68 | ||
| XM_022424914.1 | XP_022280622.1 | 2 - 3 | 120 | 68 | ||
| XM_022424915.1 | XP_022280623.1 | 2 - 3 | 116 | 68 | ||
| XM_022424916.1 | XP_022280624.1 | 2 - 3 | 120 | 68 | ||
| XM_022424917.1 | XP_022280625.1 | 2 - 3 | 107 | 68 | ||
More Information
Set Membership: |
|

PALM2AKAP2
Genomic Map