Assay Details
| Gene Symbol: PALM2AKAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204899.1 | NP_001191828.1 | 4 - 5 | 367 | 100 | |
| XM_022424914.1 | XP_022280622.1 | 5 - 6 | 448 | 100 | ||
| XM_022424915.1 | XP_022280623.1 | 5 - 6 | 444 | 100 | ||
| XM_022424916.1 | XP_022280624.1 | 5 - 6 | 448 | 100 | ||
| XM_022424917.1 | XP_022280625.1 | 5 - 6 | 435 | 100 | ||
More Information
Set Membership: |
|

PALM2AKAP2
Genomic Map