Assay Details
| Gene Symbol: IL11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022427547.1 | XP_022283255.1 | 2 - 3 | 363 | 76 | |
| XM_022427551.1 | XP_022283259.1 | 2 - 3 | 488 | 76 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_022427551.1 transcripts.
Set Membership: |
|

IL11
Genomic Map