Assay Details
| Gene Symbol: MIR764 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_049524.1 | 1 | 48 | 59 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | angiomotin | AMOT | -1.5mb | Anti-sense | Chr. X - 85742610 - 85807557 on NCBI Build CanFam3.1 | |
| Downstream | 5-hydroxytryptamine receptor 2C | HTR2C |
5-HT-2C, 5-HTR2C |
48.4kb | Sense | Chr. X - 87342800 - 87641224 on NCBI Build CanFam3.1 |
More Information
Set Membership: |
|

MIR764
Genomic Map