Assay Details
| Gene Symbol: MIR105B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_049470.1 | 1 | 38 | 74 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | gamma-aminobutyric acid type A receptor subunit alpha3 | GABRA3 | Sense | Chr. X - 120152650 - 120460604 on NCBI Build CanFam3.1 | ||
| Downstream | microRNA mir-105a | MIR105A | cfa-mir-105a | 93.3kb | Sense | Chr. X - 120403195 - 120403275 on NCBI Build CanFam3.1 |
More Information
Set Membership: |
|

MIR105B
Genomic Map