Assay Details
| Gene Symbol: FAM32A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287071.1 | NP_001274000.1 | 2 - 3 | 229 | 105 | |
| NM_001287072.1 | NP_001274001.1 | 2 - 3 | 229 | 105 | ||
| GenBank mRNA | DN269365.1 | - | 2 - 3 | 229 | 105 | |
| DN417984.1 | - | 1 | 224 | 105 | ||
More Information
Set Membership: |
|

FAM32A
Genomic Map