Assay Details
| Gene Symbol: Nupl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003500503.2 | XP_003500551.1 | 14 - 15 | 1781 | 75 | |
| XM_007641270.1 | XP_007639460.1 | 13 - 14 | 1642 | 75 | ||
| GenBank mRNA | JI880492.1 | - | 13 - 14 | 1608 | 75 | |
| JP058453.1 | - | 12 - 13 | 1111 | 75 | ||
More Information
Set Membership: |
|

Nupl1
Genomic Map