Assay Details
| Gene Symbol: Exosc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007645890.1 | XP_007644080.1 | 4 - 5 | 503 | 95 | |
| GenBank mRNA | JP037161.1 | - | 4 - 5 | 503 | 95 | |
More Information
Set Membership: |
|

Exosc7
Genomic Map