Assay Details
| Gene Symbol: Znf652 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003503944.2 | XP_003503992.1 | 4 - 5 | 1664 | 70 | |
| GenBank mRNA | JI886191.1 | - | 3 - 4 | 291 | 70 | |
| JP048343.1 | - | 3 - 4 | 291 | 70 | ||
More Information
Set Membership: |
|

Znf652
Genomic Map