Assay Details
| Gene Symbol: Kbtbd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003504900.2 | XP_003504948.1 | 2 - 3 | 504 | 69 | |
| GenBank mRNA | JI872608.1 | - | 1 - 2 | 521 | 69 | |
| JP065993.1 | - | 1 - 2 | 521 | 69 | ||
More Information
Set Membership: |
|

Kbtbd2
Genomic Map