Assay Details
| Gene Symbol: Csrp2bp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003496467.2 | XP_003496515.2 | 1 - 2 | 281 | 84 | |
| GenBank mRNA | JI894900.1 | - | 1 - 2 | 56 | 84 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Csrp2bp| Gene Symbol: Csrp2bp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003496467.2 | XP_003496515.2 | 1 - 2 | 281 | 84 | |
| GenBank mRNA | JI894900.1 | - | 1 - 2 | 56 | 84 | |