Assay Details
| Gene Symbol: Arhgap5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003512675.2 | XP_003512723.1 | 5 - 6 | 4378 | 134 | |
| GenBank mRNA | JI894597.1 | - | 5 - 6 | 3398 | 134 | |
More Information
Set Membership: |
|

Arhgap5
Genomic Map