Assay Details
| Gene Symbol: Isl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498392.2 | XP_003498440.1 | 5 - 6 | 984 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Isl1| Gene Symbol: Isl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498392.2 | XP_003498440.1 | 5 - 6 | 984 | 65 | |
Set Membership: |
|