Assay Details
| Gene Symbol: Map3k10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007645767.1 | XP_007643957.1 | 5 - 6 | 914 | 61 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Map3k10| Gene Symbol: Map3k10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007645767.1 | XP_007643957.1 | 5 - 6 | 914 | 61 | |
Set Membership: |
|