Assay Details
| Gene Symbol: Layn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001246681.1 | NP_001233610.1 | 2 - 3 | 498 | 62 | |
| XM_007629041.1 | XP_007627231.1 | 3 - 4 | 664 | 62 | ||
| GenBank mRNA | AF093673.1 | - | 2 - 3 | 522 | 62 | |
| JI889967.1 | - | 2 - 3 | 502 | 62 | ||
| JP045844.1 | - | 2 - 3 | 502 | 62 | ||
More Information
Set Membership: |
|

Layn
Genomic Map