Assay Details
| Gene Symbol: Znf330 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244298.1 | NP_001231227.1 | 4 - 5 | 439 | 65 | |
| GenBank mRNA | JI881202.1 | - | 1 - 2 | 184 | 65 | |
| JP065055.1 | - | 1 - 2 | 184 | 65 | ||
| Y12836.1 | - | 4 - 5 | 439 | 65 | ||
More Information
Set Membership: |
|

Znf330
Genomic Map