Assay Details
| Gene Symbol: Slc35a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001246818.2 | NP_001233747.1 | 5 - 6 | 765 | 88 | |
| XM_007653523.1 | XP_007651713.1 | 6 - 7 | 847 | 88 | ||
| GenBank mRNA | FN825777.1 | - | 5 - 6 | 768 | 88 | |
More Information
Set Membership: |
|

Slc35a3
Genomic Map