Assay Details
| Gene Symbol: Selt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001256851.1 | NP_001243780.1 | 4 - 5 | 547 | 79 | |
| GenBank mRNA | JI864277.1 | - | 4 - 5 | 519 | 79 | |
| JP065339.1 | - | 4 - 5 | 519 | 79 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Selt| Gene Symbol: Selt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001256851.1 | NP_001243780.1 | 4 - 5 | 547 | 79 | |
| GenBank mRNA | JI864277.1 | - | 4 - 5 | 519 | 79 | |
| JP065339.1 | - | 4 - 5 | 519 | 79 | ||