Assay Details
| Gene Symbol: Dcun1d2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003504082.2 | XP_003504130.1 | 2 - 3 | 283 | 76 | |
| XM_007644597.1 | XP_007642787.1 | 2 - 3 | 283 | 76 | ||
| XM_007644598.1 | XP_007642788.1 | 2 - 3 | 283 | 76 | ||
| XM_007644599.1 | XP_007642789.1 | 2 - 3 | 283 | 76 | ||
| XM_007644600.1 | XP_007642790.1 | 2 - 3 | 283 | 76 | ||
| GenBank mRNA | JI881115.1 | - | 2 - 3 | 249 | 76 | |
More Information
Set Membership: |
|

Dcun1d2
Genomic Map