Assay Details
| Gene Symbol: Sh3rf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003503277.2 | XP_003503325.1 | 3 - 4 | 1163 | 62 | |
| XM_007643880.1 | XP_007642070.1 | 3 - 4 | 980 | 62 | ||
| XM_007643881.1 | XP_007642071.1 | 3 - 4 | 880 | 62 | ||
| XM_007643882.1 | XP_007642072.1 | 3 - 4 | 845 | 62 | ||
More Information
Set Membership: |
|

Sh3rf2
Genomic Map