Assay Details
| Gene Symbol: Lage3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003512285.2 | XP_003512333.2 | 382 | 88 | ||
| GenBank mRNA | JP039383.1 | - | 369 | 88 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lage3| Gene Symbol: Lage3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003512285.2 | XP_003512333.2 | 382 | 88 | ||
| GenBank mRNA | JP039383.1 | - | 369 | 88 | ||
Set Membership: |
|